Anti-VTI1B Goat Polyclonal Antibody

Lieferant: ProSci Inc.
PRSI46-590EA 420 EUR
PRSI46-590
Anti-VTI1B Goat Polyclonal Antibody
Antikörper
ELISA: Antibody detection limit dilution 1:32,000. Western Blot: Approximately 26-28 kDa band observed in human duodenum, kidney and testis lysates (calculated MW of 26.7 kDa according to NP_006361.1). Recommended concentration: 1-3 μg/mL.

VTI1B antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

VTI1B antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: VTI1B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR