Anti-VPS11 Goat Polyclonal Antibody

Lieferant: ProSci Inc.
PRSI46-575EA 420 EUR
PRSI46-575
Anti-VPS11 Goat Polyclonal Antibody
Antikörper
ELISA: Antibody detection limit dilution 1:32,000. Western Blot: Approximately 75 kDa band in human duodenum, colon and placenta lysates after 0.3 μg/mL antibody staining.3. The 75 kDa band was successfully blocked by incubation with the immunizing peptide.

VPS11 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

VPS11 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: VPS11
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR