Anti-GC Goat Polyclonal Antibody

Lieferant: ProSci Inc.
PRSI46-573EA 420 EUR
PRSI46-573
Anti-GC Goat Polyclonal Antibody
Antikörper
ELISA: Antibody detection limit dilution 1:2000. Western Blot: Approximately 56 kDa band observed in human lung lysates (calculated MW of 52.9 kDa according to NP_000574.2). The observed molecular weight corresponds to earlier findings in literature with different antibodies (DiMartino and Kew, J Immunol. 1999 Aug 15;163 (4) :2135-42; PMID: 10438954). Recommended concentration: 0.1-0.3 μg/mL.

Vitamin D-binding Protein antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

Vitamin D-binding Protein antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: GC
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR