Anti-USP11 Goat Polyclonal Antibody

Lieferant: ProSci Inc.
PRSI46-555EA 553 EUR
PRSI46-555
Anti-USP11 Goat Polyclonal Antibody
Antikörper
ELISA: Antibody detection limit dilution 1:8000. Western Blot: Approximately 120 kDa band observed in human testis lysates and in Jurkat lysate (calculated MW of 110 kDa according to NP_004642.2). The observed molecular weight corresponds to earlier findings in literature with different antibodies (Ideguchi et al, Biochem J. 2002 Oct 1;367 (Pt 1) :87-95.; PMID: 12084015). Recommended concentration: 1-3 μg/mL.

USP11 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

USP11 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: USP11
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR