Anti-USH1C Goat Polyclonal Antibody

Lieferant: ProSci Inc.
PRSI46-554EA 553 EUR
PRSI46-554
Anti-USH1C Goat Polyclonal Antibody
Antikörper
ELISA: Antibody detection limit dilution 1:128,000. Western Blot: Approximately 75 kDa band observed in HEK293 lysate (calculated MW of 62.2 kDa according to NP_005700.2). Recommended concentration: 0.1-0.3 μg/mL.

USH1C antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

USH1C antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: USH1C
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR