Anti-UNC13D Goat Polyclonal Antibody

Lieferant: ProSci Inc.
PRSI46-552EA 420 EUR
PRSI46-552
Anti-UNC13D Goat Polyclonal Antibody
Antikörper
ELISA: Antibody detection limit dilution 1:128,000. Western Blot: Approximately 105 kDa band observed in human T-lymphocyte and HeLa lysates (calculated MW of 123 kDa according to NP_954712.1). Recommended concentration: 1-3 μg/mL. Immunohistochemisty: In paraffin embedded human thymus shows cytoplasm staining in select areas. Recommended concentration, 2.5 μg/mL.

UNC13D antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

UNC13D antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: UNC13D
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR