Anti-TSPO Goat Polyclonal Antibody

Lieferant: ProSci Inc.
PRSI46-538EA 420 EUR
PRSI46-538
Anti-TSPO Goat Polyclonal Antibody
Antikörper
ELISA: Antibody detection limit dilution 1:128,000. Approximately 35 kDa band observed in human brain lysates (calculated MW of 18.8 kDa according to NP_000705.2). The observed molecular weight corresponds to earlier findings in literature with different antibodies (Delavoie et al, Biochemistry. 2003 Apr 22;42 (15) :4506-19.; PMID: 12693947). Recommended concentration: 0.1-0.3 μg/mL. Approximately 17 kDa band observed in human breast carcinoma lysates (calculated MW of 18.8 kDa according to NP_000705.2). Recommended concentration: 0.1-0.3 μg/mL.

TSPO antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

TSPO antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: TSPO
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR