Anti-TSC1 Goat Polyclonal Antibody

Lieferant: ProSci Inc.
PRSI46-535EA 420 EUR
PRSI46-535
Anti-TSC1 Goat Polyclonal Antibody
Antikörper
ELISA: Antibody detection limit dilution 1:128000. Western Blot: Approximately 150 kDa band observed in human brain lysates (calculated MW of 130 kDa according to NP_000359.1). The observed molecular weight corresponds to earlier findings in literature with different antibodies (Miloloza et al Hum Mol Genet. 2000 Jul 22;9 (12) :1721-7; PMID: 10915759). In transfected HEK293 transiently expressing human TSC1 a band of approximately 120 kDa is observed. This band is not observed in the non-transfected HEK293. Recommended concentration: 0.1-0.3 μg/mL.

TSC1 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

TSC1 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: TSC1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR