Anti-TRPM7 Goat Polyclonal Antibody

Lieferant: ProSci Inc.
PRSI46-531EA 553 EUR
PRSI46-531
Anti-TRPM7 Goat Polyclonal Antibody
Antikörper
ELISA: Antibody detection limit dilution 1:16,000. Western Blot: Approximately 150 kDa band observed in human cerebellum lysates (calculated MW of 213 kDa according to NP_060142.2). The observed molecular weight corresponds to earlier findings in literature with different antibodies (Clark et al PLoS ONE. 2008 Mar 26;3 (3) :e1876.; PMID: 18365021). Recommended concentration: 0.3- 1 μg/mL. Immunohistochemistry: In paraffin embedded mouse brain shows neuronal staining in the hippocampus. Recommended concentration, 4-6 μg/mL.

TRPM7 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

TRPM7 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: TRPM7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR