Anti-TPI1 Goat Polyclonal Antibody

Lieferant: ProSci Inc.
PRSI46-528EA 420 EUR
PRSI46-528
Anti-TPI1 Goat Polyclonal Antibody
Antikörper
ELISA: Antibody detection limit dilution 1:16,000. Western Blot: Approximately 26 kDa band observed in human liver and human hepatoblastoma HepG2 cell lysates (calculated MW of 26.7 kDa according to NP_000356). Recommended concentration: 0.01-0.03 μg/mL.

Triosephosphate isomerase antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

TriosepH ospH ate isomerase antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: TPI1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR