Anti-TMEM37 Goat Polyclonal Antibody

Lieferant: ProSci Inc.
PRSI46-511EA 435 EUR
PRSI46-511
Anti-TMEM37 Goat Polyclonal Antibody
Antikörper
ELISA: Antibody detection limit dilution 1:32,000. Western Blot: Approximately 26-28 kDa band in human brain (cerebellum, frontal cortex and hippocampus) lysates after 0.1 μg/mL antibody staining.9 kDa according to NP_899063.2. The 26-28 kDa band was successfully blocked by incubation with the immunizing peptide.

Transmembrane Protein 37 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

Transmembrane Protein 37 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: TMEM37
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR