Anti-TRAF2 Goat Polyclonal Antibody

Lieferant: ProSci Inc.
PRSI46-510EA 553 EUR
PRSI46-510
Anti-TRAF2 Goat Polyclonal Antibody
Antikörper
ELISA: Antibody detection limit dilution 1:128000. Western Blot: Approximately 50 kDa band observed in human ovary lysates (calculated MW of 55.9 kDa according to NP_066961.2). In transfected HEK293 transiently expressing human TRAF2 a band of approximately 60 kDa is observed. This band is not observed in the non-transfected HEK293. The endogenous 48 kDa protein is observed in both. Recommended concentration: 0.1-0.3 μg/mL. Approximately 50 kDa band observed in human Ovary lysates (calculated MW of 55.9 kDa according to NP_066961.2). In transfected HEK293 transiently expressing human TRAF2 a band of approximately. 60 kDa is observed. This band is not observed in the non-transfected HEK293. The endogenous 48 kDa protein is observed in both. Recommended concentration: 0.1-0.3 μg/mL. Immunohistochemistry: In paraffin embedded human kidney shows cytoplasm and select nuclear staining. Recommended concentration, 3-6 μg/mL.

TRAF2 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

TRAF2 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: TRAF2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR