Anti-TRAF1 Goat Polyclonal Antibody

Lieferant: ProSci Inc.
PRSI46-509EA 420 EUR
PRSI46-509
Anti-TRAF1 Goat Polyclonal Antibody
Antikörper
ELISA: Antibody detection limit dilution 1:16,000. Western Blot: Approximately 50-55 kDa band observed in lysates of monocytic leukemia cell line U937 (calculated MW of 46.2 kDa according to NP_005649.1). The observed molecular weight corresponds to earlier findings in literature with different antibodies (Pryhuber et al, Am J Respir Cell Mol Biol. 2000 Feb;22 (2) :150-6. PMID: 10657935). Recommended concentration: 0.3-1.0 μg/mL.

TRAF1 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

TRAF1 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: TRAF1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR