Anti-TMED10 Goat Polyclonal Antibody

Lieferant: ProSci Inc.
PRSI46-502EA 553 EUR
PRSI46-502
Anti-TMED10 Goat Polyclonal Antibody
Antikörper
ELISA: Antibody detection limit dilution 1:1,000. Western Blot: Approximately 75 kDa band in human and rat pancreas lysates after 1 μg/mL antibody staining.The 75 kDa band was successfully blocked by incubation with the immunizing peptide.

TMP21 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

TMP21 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: TMED10
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR