Anti-TIMM50 Goat Polyclonal Antibody

Lieferant: ProSci Inc.
PRSI46-496EA 553 EUR
PRSI46-496
Anti-TIMM50 Goat Polyclonal Antibody
Antikörper
ELISA: Antibody detection limit dilution 1:16,000. Western Blot: experiments gave bands at approximately 40 kDa in human heart lysate after 0.5 μg/mL antibody staining. This band corresponds to earlier findings in literature with different antibodies (PMID: 15044455). This protein has a calculated MW of 50.5 kDa according to NP_001001563. Recommended concentration: 0.5-3 μg/mL.

TIM-50 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

TIM-50 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: TIMM50
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR