Anti-TRIM24 Goat Polyclonal Antibody

Lieferant: ProSci Inc.
PRSI46-493EA 420 EUR
PRSI46-493
Anti-TRIM24 Goat Polyclonal Antibody
Antikörper
ELISA: Antibody detection limit dilution 1:32,000. Western Blot: Approximately 120 kDa band in human kidney and heart lysates at 1 μg/mL, however prominent bands were also seen at 25 kDa and 45 kDa . Immunohistochemistry: In paraffin embedded human breast carcinoma shows nuclear staining in the distorted lobules. Recommended concentration, 4-6μg/ml

TIF1 alpha antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

TIF1 alpH a antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: TRIM24
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR