Anti-TMSB15A Goat Polyclonal Antibody

Lieferant: ProSci Inc.
PRSI46-490EA 345 EUR
PRSI46-490
Anti-TMSB15A Goat Polyclonal Antibody
Antikörper
ELISA: Antibody detection limit dilution 1:2000. Western Blot: No signal obtained yet but low background observed in 293 and A549 lysates at up to 2 μg/mL (please note that our SDS PAGE/Western Blotting protocol is not optimized for detection of proteins smaller than 15 kDa ).

Thymosin-like 8 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

Thymosin-like 8 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: TMSB15A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR