Anti-MAL Goat Polyclonal Antibody

Lieferant: ProSci Inc.
PRSI46-473EA 435 EUR
PRSI46-473
Anti-MAL Goat Polyclonal Antibody
Antikörper
ELISA: Antibody detection limit dilution 1:4,000. Western Blot: Approximately 35 kDa band in human thymus and thyroid gland lysates after 0.3 μg/mL antibody staining.7 kDa according to NP_002362.1. The 35 kDa band was successfully blocked by incubation with the immunizing peptide.

T-cell Differentiation Protein antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

T-cell Differentiation Protein antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: MAL
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR