Anti-MAP3K7 Goat Polyclonal Antibody

Lieferant: ProSci Inc.
PRSI46-469EA 420 EUR
PRSI46-469
Anti-MAP3K7 Goat Polyclonal Antibody
Antikörper
ELISA: Antibody detection limit dilution 1:8000. Western Blot: In transfected HEK293 transiently expressing full-length human MAP3K7 (myc and DYKDDDDK tagged), a band of approximately 75 kDa was observed. No bands were observed in mock-transfected HEK293 and the same band was observed using anti-DYKDDDDK tag antibody. Recommended concentration, 1-2 μg/mL.

TAK1 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

TAK1 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: MAP3K7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR