Anti-SUPT16H Goat Polyclonal Antibody

Lieferant: ProSci Inc.
PRSI46-450EA 553 EUR
PRSI46-450
Anti-SUPT16H Goat Polyclonal Antibody
Antikörper
ELISA: Antibody detection limit dilution 1:8,000. Western Blot: Approximately 150 kDa band observed in HeLa lysate (calculated MW of 120 kDa according to NP_009123.1). Recommended concentration: 0.1-0.3 μg/mL. an additional band of unknown identity was also consistently observed at 30 kDa . This band was successfully blocked by incubation with the immunizing peptide.

SUPT16H antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

SUPT16H antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: SUPT16H
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR