Anti-STELLAR Goat Polyclonal Antibody

Lieferant: ProSci Inc.
PRSI46-444EA 345 EUR
PRSI46-444
Anti-STELLAR Goat Polyclonal Antibody
Antikörper
ELISA: Antibody detection limit dilution 1:64,000. Western Blot: Approximately 26 kDa band in human ovary and placenta lysates after 0.1 μg/mL antibody staining. The 26 kDa band was successfully blocked by incubation with the immunizing peptide.

STELLAR antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

STELLAR antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: STELLAR
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR