Anti-SCD Goat Polyclonal Antibody

Lieferant: ProSci Inc.
PRSI46-443EA 420 EUR
PRSI46-443
Anti-SCD Goat Polyclonal Antibody
Antikörper
ELISA: Antibody detection limit dilution 1:64,000. Western Blot: Approximately 30 kDa band in human adipose, brain and liver lysates after 0.3 μg/mL antibody staining.5 kDa according to NP_005054.3. The 30 kDa band was successfully blocked by incubation with the immunizing peptide. Immunohistochemistry: In paraffin embedded human liver shows textured cytoplam staining in hepatocytes. Recommended concentration, 5-10 μg/mL.

Stearoyl-CoA desaturase antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

Stearoyl-CoA desaturase antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: SCD
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR