Anti-SPRY1 Goat Polyclonal Antibody

Lieferant: ProSci Inc.
PRSI46-430EA 553 EUR
PRSI46-430
Anti-SPRY1 Goat Polyclonal Antibody
Antikörper
ELISA: Antibody detection limit dilution 1:16000. Western Blot: this antibody has been successfully used in ERMS cells (PMID: 20068162). Recommended concentration: 1-3 μg/mL. Immunohistochemistry: this antibody has been successfully used by anonymous customers in paraffin embedded mouse brain, showing nuclear staining. Recommended concentration: 1-3 μg/mL.

Sprouty antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

Sprouty antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: SPRY1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR