Anti-SPEN Goat Polyclonal Antibody

Lieferant: ProSci Inc.
PRSI46-426EA 345 EUR
PRSI46-426
Anti-SPEN Goat Polyclonal Antibody
Antikörper
ELISA: Antibody detection limit dilution 1:4,000. Western Blot: Not yet tested - our routinely used Western Blotting protocol does not allow detection of proteins as large as the calculated size of approximately 402 kDa according to NP_055816.2. Therefore we cannot recommend an optimal concentration and the product is investigative grade.

SPEN antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

SPEN antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: SPEN
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR