Anti-SORBS1 Goat Polyclonal Antibody

Lieferant: ProSci Inc.
PRSI46-416EA 420 EUR
PRSI46-416
Anti-SORBS1 Goat Polyclonal Antibody
Antikörper
ELISA: Antibody detection limit dilution 1:32,000. Western Blot: Approximately 65 kDa band in human adipose lysates after 0.3 μg/mL antibody staining.6 kDa according to NP_006425.2 and higher sizes for other isoforms. The 65 kDa band was successfully blocked by incubation with the immunizing peptide.

SORBS1 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

SORBS1 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: SORBS1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR