Anti-SOD1 Goat Polyclonal Antibody

Lieferant: ProSci Inc.
PRSI46-412EA 420 EUR
PRSI46-412
Anti-SOD1 Goat Polyclonal Antibody
Antikörper
ELISA: Antibody detection limit dilution 1:8,000. Western Blot: Approximately 18 kDa band observed in mouse brain and rat spinal cord lysates (calculated MW of 15.9 kDa according to human NP_000445.1, mouse NP_035564.1 and rat NP_058746.1). Recommended concentration: 0.01-0.03 μg/mL.

SOD1 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

SOD1 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: SOD1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR