Anti-SMO Goat Polyclonal Antibody

Lieferant: ProSci Inc.
PRSI46-389EA 350 EUR
PRSI46-389
Anti-SMO Goat Polyclonal Antibody
Antikörper
ELISA: Antibody detection limit dilution 1:1,000. Approximately 60 kDa band observed in human bone marrow lysates (calculated MW of 86.4 kDa according to NP_005622.1). The observed molecular weight corresponds to findings with 46-388, a product of different design reacting to a different part of the same protein. Recommended concentration: 0.1-0.3 μg/mL.

SMO antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

SMO antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: SMO
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR