Anti-HLTF Goat Polyclonal Antibody

Lieferant: ProSci Inc.
PRSI46-385EA 553 EUR
PRSI46-385
Anti-HLTF Goat Polyclonal Antibody
Antikörper
ELISA: Antibody detection limit dilution 1:16000. Western Blot: Approximately 120 kDa and 50 kDa bands seen in Jurkat cell lysate. Recommended for use at 0.2-1 μg/mL. Please note that the weaker smaller band may represent a novel isoform of this protein and is in agreement with previous Western Blot observations (J Biol Chem 1995 Mar 3;270 (9) :4575-87).

SMARCA3 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

SMARCA3 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: HLTF
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR