Anti-SMAD9 Goat Polyclonal Antibody

Lieferant: ProSci Inc.
PRSI46-383EA 553 EUR
PRSI46-383
Anti-SMAD9 Goat Polyclonal Antibody
Antikörper
ELISA: Antibody detection limit dilution 1:64,000. Western Blot: Approximately 50 kDa band observed in human skin lysates (calculated MW of 48.6 kDa according to NP_005896.1). Recommended concentration: 0.3-1.0 μg/mL.

Smad9 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

Smad9 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: SMAD9
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR