Anti-SLC22A16 Goat Polyclonal Antibody

Lieferant: ProSci Inc.
PRSI46-375EA 553 EUR
PRSI46-375
Anti-SLC22A16 Goat Polyclonal Antibody
Antikörper
ELISA: Antibody detection limit dilution 1:128,000. Western Blot: Approximately 85-90 kDa band in human testis lysates after 0.5 μg/mL antibody staining.6 kDa according to NP_149116.2. The 85-90 kDa band was successfully blocked by incubation with the immunizing peptide.

SLC22A16 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

SLC22A16 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: SLC22A16
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR