Anti-SIRT3 Goat Polyclonal Antibody

Lieferant: ProSci Inc.
PRSI46-372EA 420 EUR
PRSI46-372
Anti-SIRT3 Goat Polyclonal Antibody
Antikörper
ELISA: Antibody detection limit dilution 1:128000. Western Blot: Approximately 28 kDa band observed in human liver lysates (calculated MW of 28.6 kDa according to NP_001017524.1). In transfected HEK293 transiently expressing SIRT3 bands of approximately 35 kDa and 48 kDa are observed. These bands are not observed in the non-transfected HEK293. Recommended concentration: 0.1-0.3 μg/mL.

SIRT3 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

SIRT3 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: SIRT3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR