Anti-SH3BP1 Goat Polyclonal Antibody

Lieferant: ProSci Inc.
PRSI46-367EA 420 EUR
PRSI46-367
Anti-SH3BP1 Goat Polyclonal Antibody
Antikörper
ELISA: Antibody detection limit dilution 1:16000. Western Blot: Approximately 70 kDa band observed in human lung and kidney lysates (predicted MW of 71 kDa according to NP_061830). Recommended for use at 1-3 μg/mL. Immunohistochemistry: In paraffin embedded human kidney, stains the cytoplasm of lymphoid cells in the interstitial parenchyma and of lymphocytes in a glomerulus. Recommended concentration, 0.3 μg/mL.

SH3BP1 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

SH3BP1 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: SH3BP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR