Anti-SH2D1A Goat Polyclonal Antibody

Lieferant: ProSci Inc.
PRSI46-365EA 420 EUR
PRSI46-365
Anti-SH2D1A Goat Polyclonal Antibody
Antikörper
ELISA: Antibody detection limit dilution 1:16000. Western Blot: Approximately 16 kDa band observed in human thymus lysates (calculated MW of 14.2x kDa according to NP_002342.1). In transfected HEK293 transiently expressing SH2D1A a band of approximately 16 kDa is observed. This band is not observed in the non-transfected HEK293. Recommended concentration: 0.3-1 μg/mL.

SH2D1A antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

SH2D1A antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: SH2D1A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR