Anti-HTR2C Goat Polyclonal Antibody

Lieferant: ProSci Inc.
PRSI46-356EA 420 EUR
PRSI46-356
Anti-HTR2C Goat Polyclonal Antibody
Antikörper
ELISA: Antibody detection limit dilution 1:64,000. Western Blot: Approximately 50 kDa band observed in lysates of human EBV immortalised lymphoblastoids, T cell MOLT4 and monocytic cell line U937 (calculated MW of 51.8 kDa according to NP_000859.1). Recommended concentration: 1-3 μg/mL.

Serotonin receptor 2C antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

Serotonin receptor 2C antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: HTR2C
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR