Anti-HTR1B Goat Polyclonal Antibody

Lieferant: ProSci Inc.
PRSI46-355EA 420 EUR
PRSI46-355
Anti-HTR1B Goat Polyclonal Antibody
Antikörper
ELISA: Antibody detection limit dilution 1:8,000. Western Blot: Approximately 48 kDa band observed in human brain lysates (calculated MW of 43.6 kDa according to NP_000854.1). Recommended concentration: 1-3 μg/mL.

Serotonin receptor 1B antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

Serotonin receptor 1B antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: HTR1B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR