Anti-SEPT1 Goat Polyclonal Antibody

Lieferant: ProSci Inc.
PRSI46-352EA 420 EUR
PRSI46-352
Anti-SEPT1 Goat Polyclonal Antibody
Antikörper
ELISA: Antibody detection limit dilution 1:32,000. Western Blot: Approximately 30 kDa band in mouse and rat brain lysates after 1 μg/mL antibody staining.0 kDa according to mouse NP_059489.1. The 30 kDa band was successfully blocked by incubation with the immunizing peptide. Approximately 45 kDa band observed in Jurkat lysate (calculated MW of 42.0 kDa according to NP_443070.1). Recommended concentration: 2-4 μg/mL.

Septin 1 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

Septin 1 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: SEPT1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR