Anti-SENP7 Goat Polyclonal Antibody

Lieferant: ProSci Inc.
PRSI46-351EA 345 EUR
PRSI46-351
Anti-SENP7 Goat Polyclonal Antibody
Antikörper
ELISA: Antibody detection limit dilution 1:16,000. Western Blot: Approximately 90 kDa band in human cerebellum and thymus lysates after 0.3 μg/mL antibody staining.1 and of 120 kDa according to NP_065705.3. The 90 kDa band was successfully blocked by incubation with the immunizing peptide.

SENP7 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

SENP7 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: SENP7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR