Anti-SLFN8 Goat Polyclonal Antibody

Lieferant: ProSci Inc.
PRSI46-342EA 325 EUR
PRSI46-342
Anti-SLFN8 Goat Polyclonal Antibody
Antikörper
ELISA: Antibody detection limit dilution 1:64,000. Western Blot: Approximately 70 kDa band in mouse liver lysates after 1 μg/mL antibody staining. The 70 kDa band was successfully blocked by incubation with the immunizing peptide.

Schlafen 8 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

Schlafen 8 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: SLFN8
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR