Anti-SECISBP2 Goat Polyclonal Antibody

Lieferant: ProSci Inc.
PRSI46-339EA 420 EUR
PRSI46-339
Anti-SECISBP2 Goat Polyclonal Antibody
Antikörper
ELISA: Antibody detection limit dilution 1:64000. Western Blot: Approximately 120 kDa band detected in HFF cell lysates. Data kindly provided by Laura Papp and Kum Kum Khanna. The observed molecular weight corresponds to earlier findings in literature with different antibodies (Papp et al, Mol Cell Biol. 2006 Jul;26 (13) :4895-910; PMID: 16782878). Recommended concentration: 0.1-0.3 μg/mL. Immunohistochemistry: In paraffin embedded human testis shows granular cytoplasm staining in Leydig cells. Recommended concentration, 3 μg/mL.

SBP2 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

SBP2 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: SECISBP2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR