Anti-SALL1 Goat Polyclonal Antibody

Lieferant: ProSci Inc.
PRSI46-332EA 345 EUR
PRSI46-332
Anti-SALL1 Goat Polyclonal Antibody
Antikörper
ELISA: Antibody detection limit dilution 1:1,000. Western Blot: Approximately 40 kDa band in human umbilical cord, ovary, placenta and testis lysates after 0.1 μg/mL antibody staining. The 40 kDa band was successfully blocked by incubation with the immunizing peptide.

SALL1 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

SALL1 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: SALL1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR