Anti-ROBO3 Goat Polyclonal Antibody

Lieferant: ProSci Inc.
PRSI46-319EA 345 EUR
PRSI46-319
Anti-ROBO3 Goat Polyclonal Antibody
Antikörper
ELISA: Antibody detection limit dilution 1:16,000. Western Blot: Approximately 75-80 kDa band in mouse brain and rat brain lysates after 0.1 μg/mL antibody staining.1and AAD11628.1. The 75-80 kDa band was successfully blocked by incubation with the immunizing peptide.

Robo3 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

Robo3 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: ROBO3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR