Anti-ROBO1 Goat Polyclonal Antibody

Lieferant: ProSci Inc.
PRSI46-316EA 420 EUR
PRSI46-316
Anti-ROBO1 Goat Polyclonal Antibody
Antikörper
ELISA: Antibody detection limit dilution 1:128,000. Western Blot: Approximately 80 kDa and 40 kDa in lysates of mouse brain and rat brain after 0.03 μg/mL antibody staining.1. Both detected bands were successfully blocked by incubation with the immunizing peptide (and BLAST results with the immunizing peptide sequence did not identify any other proteins to explain the additional bands).

Robo1 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

Robo1 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: ROBO1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR