Anti-TRIM69 Goat Polyclonal Antibody

Lieferant: ProSci Inc.
PRSI46-311EA 420 EUR
PRSI46-311
Anti-TRIM69 Goat Polyclonal Antibody
Antikörper
ELISA: Antibody detection limit dilution 1:32,000. Western Blot: Approximately 20-25 kDa band in human testis lysates after 1 μg/mL antibody staining. Approximately 55 kDa band observed in mouse testis lysates (calculated MW of 57.3 kDa according to mouse NP_536771.2). Recommended concentration: 0.5-2 μg/mL. Immunohistochemistry: In paraffin embedded human testis shows speckled staining in nuclei of spermatocytes and spermitids. Recommended concentration, 2.5-3.8 μg/mL.

RNF36 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

RNF36 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: TRIM69
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR