Anti-TRIM39 Goat Polyclonal Antibody

Lieferant: ProSci Inc.
PRSI46-307EA 420 EUR
PRSI46-307
Anti-TRIM39 Goat Polyclonal Antibody
Antikörper
ELISA: Antibody detection limit dilution >1:32000. Western Blot: No signal obtained yet but low background observed in human kidney and A431 lysates at up to 1 μg/mL.

RNF23 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

RNF23 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: TRIM39
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR