Anti-RPL19 Goat Polyclonal Antibody

Lieferant: ProSci Inc.
PRSI46-295EA 420 EUR
PRSI46-295
Anti-RPL19 Goat Polyclonal Antibody
Antikörper
ELISA: Antibody detection limit dilution 1:64,000. Western Blot: Approximately 25 kDa band observed in human testis lysates (calculated MW of 23.5 kDa according to NP_000972.1). Recommended concentration: 0.1-0.3 μg/mL.

Ribosomal Protein L19 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

Ribosomal Protein L19 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: RPL19
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR