Anti-ZFYVE20 Goat Polyclonal Antibody

Lieferant: ProSci Inc.
PRSI46-259EA 420 EUR
PRSI46-259
Anti-ZFYVE20 Goat Polyclonal Antibody
Antikörper
ELISA: Antibody detection limit dilution 1:64,000. Western Blot: Approximately 110 kDa band observed in A431, Fibroblast (epidermoid carcinoma) lysates (calculated MW of 88.8 kDa according to NP_071735). Band migration is consistent with literature (PMID 11062261, 15020713). Recommended concentration: 0.3 - 1.5 μg/mL.

Rabenosyn 5 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

Rabenosyn 5 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: ZFYVE20
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR