Anti-RAB2A Goat Polyclonal Antibody
PRSI46-258EA 420 EUR
PRSI46-258
Anti-RAB2A Goat Polyclonal Antibody
Western Blot: Approximately 23 kDa and 18 kDa bands seen in HeLa cell lysates. Recommended for use at 1-3 μg/mL.
RAB2 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.
RAB2 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.
Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.
Type:
Antigen: RAB2A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human