Anti-PRODH Goat Polyclonal Antibody

Lieferant: ProSci Inc.
PRSI46-226EA 420 EUR
PRSI46-226
Anti-PRODH Goat Polyclonal Antibody
Antikörper
ELISA: Antibody detection limit dilution 1:16,000. Western Blot: Approximately 85 kDa and 25 kDa in human brain lysates after 1 μg/mL antibody staining.0 kDa according to NP_057419.2. Both detected bands were successfully blocked by incubation with the immunizing peptide. Immunohistochemistry In paraffin embedded cerebellum shows mitochondrial staining in smaller neurons. Recommended concentration, 3-6 μg/mL.

PRODH antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

PRODH antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: PRODH
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR