Anti-PPID Goat Polyclonal Antibody

Lieferant: ProSci Inc.
PRSI46-209EA 420 EUR
PRSI46-209
Anti-PPID Goat Polyclonal Antibody
Antikörper
ELISA: Antibody detection limit dilution 1:64,000. Western Blot: Approximately 40 kDa band observed in human frontal cortex lysates (calculated MW of 40.8 kDa according to NP_005029.1). In transfected HEK293 transiently expressing CyP-40 a band of approximately 45 kDa is observed. This band is not observed in the non-transfected HEK293. Recommended concentration: 0.1-0.3 μg/mL.

PPID antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

PPID antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: PPID
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR