Anti-POMC Goat Polyclonal Antibody

Lieferant: ProSci Inc.
PRSI46-208EA 553 EUR
PRSI46-208
Anti-POMC Goat Polyclonal Antibody
Antikörper
ELISA: Antibody detection limit dilution 1:32,000. Western Blot: Approximately 26 kDa band observed in lysates of mouse pituitary cell line NIH3T3 (calculated MW of 29.4 kDa according to NP_000930.1 and NP_001030333.1). Recommended concentration: 0.01-0.03 μg/mL. Immunohistochemistry: In paraffin embedded human adrenal gland shows strong cytoplasm staining in select cells in the medulla. Recommended concentration, 2-4 μg/mL.

POMC antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

POMC antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: POMC
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR